Assay Details
| Gene Symbol: LOC4339897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788367.2 | XP_015643853.1 | 3 - 4 | 676 | 118 | |
| GenBank mRNA | AK061381.1 | - | 3 - 4 | 517 | 118 | |
| AK069439.1 | - | 3 - 4 | 507 | 118 | ||
| AK103951.1 | - | 3 - 4 | 492 | 118 | ||
| AK104422.1 | - | 3 - 4 | 504 | 118 | ||
| AK104612.1 | - | 3 - 4 | 482 | 118 | ||
More Information
Set Membership: |
|

LOC4339897
Genomic Map