Assay Details
| Gene Symbol: LOC4339897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788367.2 | XP_015643853.1 | 4 - 5 | 868 | 84 | |
| GenBank mRNA | AK061381.1 | - | 4 - 5 | 709 | 84 | |
| AK069439.1 | - | 4 - 5 | 699 | 84 | ||
| AK103951.1 | - | 4 - 5 | 684 | 84 | ||
| AK104422.1 | - | 4 - 5 | 696 | 84 | ||
| AK104612.1 | - | 4 - 5 | 674 | 84 | ||
More Information
Set Membership: |
|

LOC4339897
Genomic Map