Assay Details
| Gene Symbol: LOC4340044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785669.2 | XP_015641155.1 | 3 - 4 | 281 | 60 | |
| GenBank mRNA | AK061385.1 | - | 3 - 4 | 233 | 60 | |
| AK099815.1 | - | 3 - 4 | 251 | 60 | ||
| AK104375.1 | - | 3 - 4 | 233 | 60 | ||
More Information
Set Membership: |
|

LOC4340044
Genomic Map