Assay Details
| Gene Symbol: LOC4340044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785669.2 | XP_015641155.1 | 4 - 5 | 393 | 116 | |
| GenBank mRNA | AK061385.1 | - | 4 - 5 | 345 | 116 | |
| AK099815.1 | - | 4 - 5 | 363 | 116 | ||
| AK104375.1 | - | 4 - 5 | 345 | 116 | ||
More Information
Set Membership: |
|

LOC4340044
Genomic Map