Assay Details
| Gene Symbol: LOC4340128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786570.2 | XP_015642056.1 | 2 - 3 | 439 | 99 | |
| GenBank mRNA | AK060175.1 | - | 2 - 3 | 420 | 99 | |
| AK073593.1 | - | 2 - 3 | 425 | 99 | ||
| AK109420.2 | - | 2 - 3 | 439 | 99 | ||
More Information
Set Membership: |
|

LOC4340128
Genomic Map