Assay Details
| Gene Symbol: LOC4340192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785450.1 | XP_015640936.1 | 3 - 4 | 585 | 98 | |
| GenBank mRNA | AK059986.1 | - | 3 - 4 | 585 | 98 | |
| AK101378.1 | - | 3 - 4 | 586 | 98 | ||
More Information
Set Membership: |
|

LOC4340192
Genomic Map