Assay Details
| Gene Symbol: LOC4340317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788597.2 | XP_015644083.2 | 7 - 8 | 1513 | 72 | |
| GenBank mRNA | AK069056.1 | - | 7 - 8 | 1098 | 72 | |
| AK105719.1 | - | 7 - 8 | 1414 | 72 | ||
More Information
Set Membership: |
|

LOC4340317
Genomic Map