Assay Details
| Gene Symbol: LOC4340317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788597.2 | XP_015644083.2 | 9 - 10 | 1791 | 131 | |
| GenBank mRNA | AK069056.1 | - | 9 - 10 | 1376 | 131 | |
| AK105719.1 | - | 9 - 10 | 1692 | 131 | ||
More Information
Set Membership: |
|

LOC4340317
Genomic Map