Assay Details
Gene Symbol: LOC9268973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787609.2 | XP_015643095.1 | 23 - 24 | 3282 | 141 | |
GenBank mRNA | AK065390.1 | - | 3 - 4 | 260 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
