Assay Details
Gene Symbol: LOC9268973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787609.2 | XP_015643095.1 | 27 - 28 | 3682 | 81 | |
GenBank mRNA | AK065390.1 | - | 7 - 8 | 660 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
