Assay Details
| Gene Symbol: LOC4340386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787539.2 | XP_015643025.1 | 11 - 12 | 1622 | 118 | |
| XM_015787540.2 | XP_015643026.1 | 11 - 12 | 1589 | 118 | ||
| XM_015787541.2 | XP_015643027.1 | 11 - 12 | 2231 | 118 | ||
| GenBank mRNA | AK072074.1 | - | 11 - 12 | 1543 | 118 | |
| AK098923.1 | - | 11 - 12 | 1506 | 118 | ||
| AK100546.1 | - | 11 - 12 | 2237 | 118 | ||
| AK119193.1 | - | 7 - 8 | 979 | 118 | ||
| EF122480.1 | - | 11 - 12 | 1488 | 118 | ||
| EF122481.1 | - | 11 - 12 | 1523 | 118 | ||
| HQ895720.1 | - | 10 - 11 | 1456 | 118 | ||
| JF969237.1 | - | 10 - 11 | 1475 | 118 | ||
More Information
Set Membership: |
|

LOC4340386
Genomic Map