Assay Details
| Gene Symbol: LOC4340386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787539.2 | XP_015643025.1 | 13 - 14 | 2167 | 59 | |
| XM_015787540.2 | XP_015643026.1 | 13 - 14 | 2134 | 59 | ||
| XM_015787541.2 | XP_015643027.1 | 13 - 14 | 2776 | 59 | ||
| GenBank mRNA | AK072074.1 | - | 13 - 14 | 2088 | 59 | |
| AK098923.1 | - | 13 - 14 | 2051 | 59 | ||
| AK100546.1 | - | 13 - 14 | 2782 | 59 | ||
| AK119193.1 | - | 9 - 10 | 1524 | 59 | ||
| BT132423.1 | - | 1 - 2 | 169 | 59 | ||
| EF122480.1 | - | 13 - 14 | 2033 | 59 | ||
| EF122481.1 | - | 13 - 14 | 2068 | 59 | ||
| HQ895720.1 | - | 12 - 13 | 2001 | 59 | ||
| JF969237.1 | - | 12 - 13 | 2020 | 59 | ||
More Information
Set Membership: |
|

LOC4340386
Genomic Map