Assay Details
| Gene Symbol: LOC4340386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787539.2 | XP_015643025.1 | 3 - 4 | 384 | 136 | |
| XM_015787540.2 | XP_015643026.1 | 3 - 4 | 351 | 136 | ||
| XM_015787541.2 | XP_015643027.1 | 3 - 4 | 993 | 136 | ||
| GenBank mRNA | AK072074.1 | - | 3 - 4 | 305 | 136 | |
| AK098923.1 | - | 3 - 4 | 268 | 136 | ||
| AK100546.1 | - | 3 - 4 | 999 | 136 | ||
| EF122480.1 | - | 3 - 4 | 250 | 136 | ||
| EF122481.1 | - | 3 - 4 | 285 | 136 | ||
| HQ895720.1 | - | 2 - 3 | 218 | 136 | ||
| JF969237.1 | - | 2 - 3 | 237 | 136 | ||
More Information
Set Membership: |
|

LOC4340386
Genomic Map