Assay Details
| Gene Symbol: LOC4340386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787539.2 | XP_015643025.1 | 7 - 8 | 1061 | 92 | |
| XM_015787540.2 | XP_015643026.1 | 7 - 8 | 1028 | 92 | ||
| XM_015787541.2 | XP_015643027.1 | 7 - 8 | 1670 | 92 | ||
| GenBank mRNA | AK072074.1 | - | 7 - 8 | 982 | 92 | |
| AK098923.1 | - | 7 - 8 | 945 | 92 | ||
| AK100546.1 | - | 7 - 8 | 1676 | 92 | ||
| AK119193.1 | - | 3 - 4 | 418 | 92 | ||
| EF122480.1 | - | 7 - 8 | 927 | 92 | ||
| EF122481.1 | - | 7 - 8 | 962 | 92 | ||
| HQ895720.1 | - | 6 - 7 | 895 | 92 | ||
| JF969237.1 | - | 6 - 7 | 914 | 92 | ||
More Information
Set Membership: |
|

LOC4340386
Genomic Map