Assay Details
| Gene Symbol: LOC4340386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787539.2 | XP_015643025.1 | 8 - 9 | 1166 | 108 | |
| XM_015787540.2 | XP_015643026.1 | 8 - 9 | 1133 | 108 | ||
| XM_015787541.2 | XP_015643027.1 | 8 - 9 | 1775 | 108 | ||
| GenBank mRNA | AK072074.1 | - | 8 - 9 | 1087 | 108 | |
| AK098923.1 | - | 8 - 9 | 1050 | 108 | ||
| AK100546.1 | - | 8 - 9 | 1781 | 108 | ||
| AK119193.1 | - | 4 - 5 | 523 | 108 | ||
| EF122480.1 | - | 8 - 9 | 1032 | 108 | ||
| EF122481.1 | - | 8 - 9 | 1067 | 108 | ||
| HQ895720.1 | - | 7 - 8 | 1000 | 108 | ||
| JF969237.1 | - | 7 - 8 | 1019 | 108 | ||
More Information
Set Membership: |
|

LOC4340386
Genomic Map