Assay Details
| Gene Symbol: LOC4341078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788086.2 | XP_015643572.1 | 1 - 2 | 4376 | 129 | |
| XM_015788087.2 | XP_015643573.1 | 2 - 3 | 4292 | 129 | ||
| GenBank mRNA | AK120595.1 | - | 1 - 2 | 4355 | 129 | |
More Information
Set Membership: |
|

LOC4341078
Genomic Map