Assay Details
| Gene Symbol: LOC4341236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785777.2 | XP_015641263.1 | 1 | 1768 | 139 | |
| GenBank mRNA | AK065374.1 | - | 1 | 1754 | 139 | |
More Information
Set Membership: |
|

LOC4341236
Genomic Map