Assay Details
| Gene Symbol: LOC4331947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777281.2 | XP_015632767.1 | 4 - 5 | 1571 | 89 | |
| GenBank mRNA | AK102651.1 | - | 4 - 5 | 987 | 89 | |
More Information
Set Membership: |
|

LOC4331947
Genomic Map