Assay Details
| Gene Symbol: LOC4334033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776041.2 | XP_015631527.1 | 3 - 4 | 961 | 59 | |
| GenBank mRNA | AK060947.1 | - | 3 - 4 | 795 | 59 | |
| AK073026.1 | - | 3 - 4 | 750 | 59 | ||
| AK104415.1 | - | 3 - 4 | 796 | 59 | ||
| AK104642.1 | - | 3 - 4 | 748 | 59 | ||
More Information
Set Membership: |
|

LOC4334033
Genomic Map