Assay Details
| Gene Symbol: LOC4341697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786224.2 | XP_015641710.1 | 3 | 2087 | 92 | |
| GenBank mRNA | AK108530.1 | - | 1 | 844 | 92 | |
More Information
Set Membership: |
|

LOC4341697
Genomic Map