Assay Details
| Gene Symbol: LOC4341796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787567.1 | XP_015643053.1 | 4 - 5 | 1992 | 133 | |
| GenBank mRNA | AK060805.1 | - | 4 - 5 | 1751 | 133 | |
| AK064913.1 | - | 4 - 5 | 1941 | 133 | ||
| AK072047.1 | - | 5 - 6 | 1691 | 133 | ||
More Information
Set Membership: |
|

LOC4341796
Genomic Map