Assay Details
| Gene Symbol: LOC4341804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789057.2 | XP_015644543.1 | 1 - 2 | 344 | 62 | |
| GenBank mRNA | AK072223.1 | - | 1 - 2 | 343 | 62 | |
| AK099323.1 | - | 1 - 2 | 343 | 62 | ||
More Information
Set Membership: |
|

LOC4341804
Genomic Map