Assay Details
| Gene Symbol: LOC4342055 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785759.1 | XP_015641245.1 | 2 - 3 | 520 | 118 | |
| GenBank mRNA | AK059671.1 | - | 2 - 3 | 332 | 118 | |
| AK101018.1 | - | 2 - 3 | 333 | 118 | ||
More Information
Set Membership: |
|

LOC4342055
Genomic Map