Assay Details
| Gene Symbol: LOC4342177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790797.2 | XP_015646283.1 | 1 - 2 | 542 | 125 | |
| GenBank mRNA | AK100327.1 | - | 1 - 2 | 534 | 125 | |
| AK100796.1 | - | 1 - 2 | 534 | 125 | ||
More Information
Set Membership: |
|

LOC4342177
Genomic Map