Assay Details
| Gene Symbol: LOC4342177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790797.2 | XP_015646283.1 | 2 - 3 | 632 | 67 | |
| GenBank mRNA | AK100327.1 | - | 2 - 3 | 624 | 67 | |
| AK100796.1 | - | 2 - 3 | 624 | 67 | ||
More Information
Set Membership: |
|

LOC4342177
Genomic Map