Assay Details
| Gene Symbol: LOC4342214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789773.2 | XP_015645259.1 | 7 - 8 | 855 | 68 | |
| XM_015789775.2 | XP_015645261.1 | 7 - 8 | 868 | 68 | ||
| GenBank mRNA | AB003325.1 | - | 7 - 8 | 798 | 68 | |
| AK072683.1 | - | 6 - 7 | 2190 | 68 | ||
| AY551920.1 | - | 7 - 8 | 841 | 68 | ||
More Information
Set Membership: |
|

LOC4342214
Genomic Map