Assay Details
| Gene Symbol: LOC4342364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792204.1 | XP_015647690.1 | 1 - 2 | 201 | 64 | |
| GenBank mRNA | AK065558.1 | - | 1 - 2 | 202 | 64 | |
| AK099140.1 | - | 1 - 2 | 176 | 64 | ||
| AK099183.1 | - | 1 - 2 | 166 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792204.1 transcripts.
Set Membership: |
|

LOC4342364
Genomic Map