Assay Details
| Gene Symbol: LOC4342364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792204.1 | XP_015647690.1 | 2 - 3 | 363 | 90 | |
| GenBank mRNA | AB096864.1 | - | 1 - 2 | 160 | 90 | |
| AK065558.1 | - | 2 - 3 | 364 | 90 | ||
| AK099140.1 | - | 2 - 3 | 338 | 90 | ||
| AK099183.1 | - | 2 - 3 | 328 | 90 | ||
More Information
Set Membership: |
|

LOC4342364
Genomic Map