Assay Details
| Gene Symbol: LOC4342364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792204.1 | XP_015647690.1 | 4 - 5 | 599 | 101 | |
| GenBank mRNA | AB096864.1 | - | 3 - 4 | 396 | 101 | |
| AK099140.1 | - | 4 - 5 | 574 | 101 | ||
More Information
Set Membership: |
|

LOC4342364
Genomic Map