Assay Details
| Gene Symbol: LOC4342364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792204.1 | XP_015647690.1 | 7 - 8 | 121 | ||
| GenBank mRNA | AB096864.1 | - | 6 - 7 | 741 | 121 | |
| AK059685.1 | - | 3 - 4 | 121 | |||
| AK065558.1 | - | 7 - 8 | 121 | |||
| AK099140.1 | - | 7 - 8 | 121 | |||
| AK099183.1 | - | 7 - 8 | 121 | |||
More Information
Set Membership: |
|

LOC4342364
Genomic Map