Assay Details
| Gene Symbol: LOC4342364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792204.1 | XP_015647690.1 | 9 - 10 | 1175 | 86 | |
| GenBank mRNA | AB096864.1 | - | 8 - 9 | 972 | 86 | |
| AK059685.1 | - | 5 - 6 | 554 | 86 | ||
| AK065558.1 | - | 9 - 10 | 1175 | 86 | ||
| AK099140.1 | - | 9 - 10 | 1150 | 86 | ||
| AK099183.1 | - | 9 - 10 | 1140 | 86 | ||
More Information
Set Membership: |
|

LOC4342364
Genomic Map