Assay Details
Gene Symbol: LOC4342503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792411.2 | XP_015647897.1 | 1 - 2 | 159 | 58 | |
GenBank mRNA | AK121813.1 | - | 1 - 2 | 143 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792411.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
