Assay Details
| Gene Symbol: LOC4342542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790069.2 | XP_015645555.1 | 13 - 14 | 956 | 77 | |
| GenBank mRNA | AK066812.1 | - | 13 - 14 | 905 | 77 | |
| AK067073.1 | - | 13 - 14 | 916 | 77 | ||
More Information
Set Membership: |
|

LOC4342542
Genomic Map