Assay Details
| Gene Symbol: LOC4342542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790069.2 | XP_015645555.1 | 1 - 2 | 194 | 80 | |
| GenBank mRNA | AK066812.1 | - | 1 - 2 | 143 | 80 | |
| AK067073.1 | - | 1 - 2 | 154 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015790069.2 transcripts.
Set Membership: |
|

LOC4342542
Genomic Map