Assay Details
| Gene Symbol: LOC4342542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790069.2 | XP_015645555.1 | 9 - 10 | 712 | 93 | |
| GenBank mRNA | AK066812.1 | - | 9 - 10 | 661 | 93 | |
| AK067073.1 | - | 9 - 10 | 672 | 93 | ||
More Information
Set Membership: |
|

LOC4342542
Genomic Map