Assay Details
| Gene Symbol: LOC4342598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791348.2 | XP_015646834.1 | 13 - 14 | 1607 | 82 | |
| XM_015791349.2 | XP_015646835.1 | 13 - 14 | 1607 | 82 | ||
| GenBank mRNA | AK065328.1 | - | 2 - 3 | 2129 | 82 | |
More Information
Set Membership: |
|

LOC4342598
Genomic Map