Assay Details
| Gene Symbol: LOC4342646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789301.2 | XP_015644787.1 | 4 - 5 | 974 | 58 | |
| GenBank mRNA | AK069901.1 | - | 4 - 5 | 926 | 58 | |
More Information
Set Membership: |
|

LOC4342646
Genomic Map