Assay Details
| Gene Symbol: LOC4342827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789170.1 | XP_015644656.1 | 1 - 2 | 154 | 85 | |
| GenBank mRNA | AK104078.1 | - | 1 - 2 | 116 | 85 | |
| AK111786.1 | - | 1 - 2 | 123 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015789170.1 transcripts.
Set Membership: |
|

LOC4342827
Genomic Map