Assay Details
| Gene Symbol: LOC4343001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792539.2 | XP_015648025.1 | 12 - 13 | 1426 | 140 | |
| XM_015792541.2 | XP_015648027.1 | 12 - 13 | 1426 | 140 | ||
| GenBank mRNA | AK065714.1 | - | 12 - 13 | 1376 | 140 | |
| AK072253.1 | - | 12 - 13 | 1389 | 140 | ||
More Information
Set Membership: |
|

LOC4343001
Genomic Map