Assay Details
| Gene Symbol: LOC4343001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792539.2 | XP_015648025.1 | 2 - 3 | 365 | 146 | |
| XM_015792541.2 | XP_015648027.1 | 2 - 3 | 365 | 146 | ||
| GenBank mRNA | AK065714.1 | - | 2 - 3 | 315 | 146 | |
| AK072253.1 | - | 2 - 3 | 328 | 146 | ||
More Information
Set Membership: |
|

LOC4343001
Genomic Map