Assay Details
| Gene Symbol: LOC4343001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792539.2 | XP_015648025.1 | 5 - 6 | 557 | 130 | |
| XM_015792541.2 | XP_015648027.1 | 5 - 6 | 557 | 130 | ||
| GenBank mRNA | AK065714.1 | - | 5 - 6 | 507 | 130 | |
| AK072253.1 | - | 5 - 6 | 520 | 130 | ||
More Information
Set Membership: |
|

LOC4343001
Genomic Map