Assay Details
| Gene Symbol: LOC4343001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792539.2 | XP_015648025.1 | 7 - 8 | 1046 | 92 | |
| XM_015792541.2 | XP_015648027.1 | 7 - 8 | 1046 | 92 | ||
| GenBank mRNA | AK072253.1 | - | 7 - 8 | 1009 | 92 | |
More Information
Set Membership: |
|

LOC4343001
Genomic Map