Assay Details
| Gene Symbol: LOC4343003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792198.2 | XP_015647684.1 | 4 - 5 | 623 | 107 | |
| XM_015792199.1 | XP_015647685.1 | 4 - 5 | 600 | 107 | ||
| GenBank mRNA | AK070846.1 | - | 4 - 5 | 602 | 107 | |
More Information
Set Membership: |
|

LOC4343003
Genomic Map