Assay Details
| Gene Symbol: LOC4343013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792246.2 | XP_015647732.1 | 10 - 11 | 1685 | 137 | |
| GenBank mRNA | AK106540.1 | - | 2 - 3 | 301 | 137 | |
More Information
Set Membership: |
|

LOC4343013
Genomic Map