Assay Details
| Gene Symbol: LOC4343013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792246.2 | XP_015647732.1 | 9 - 10 | 1429 | 151 | |
| GenBank mRNA | AK103479.1 | - | 9 - 10 | 1410 | 151 | |
| AK106540.1 | - | 1 - 2 | 45 | 151 | ||
More Information
Set Membership: |
|

LOC4343013
Genomic Map