Assay Details
| Gene Symbol: LOC4343133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791833.2 | XP_015647319.1 | 2 - 3 | 553 | 105 | |
| GenBank mRNA | AK061880.1 | - | 2 - 3 | 470 | 105 | |
| AK099046.1 | - | 2 - 3 | 468 | 105 | ||
More Information
Set Membership: |
|

LOC4343133
Genomic Map