Assay Details
| Gene Symbol: LOC4343214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790881.2 | XP_015646367.1 | 1 | 839 | 152 | |
| GenBank mRNA | AK060076.1 | - | 1 | 843 | 152 | |
More Information
Set Membership: |
|

LOC4343214
Genomic Map