Assay Details
| Gene Symbol: LOC4343296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792215.2 | XP_015647701.1 | 2 - 3 | 2443 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4343296| Gene Symbol: LOC4343296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792215.2 | XP_015647701.1 | 2 - 3 | 2443 | 108 | |
Set Membership: |
|