Assay Details
| Gene Symbol: LOC4343296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792215.2 | XP_015647701.1 | 3 - 4 | 2520 | 112 | |
| XM_015792216.2 | XP_015647702.1 | 3 - 4 | 2514 | 112 | ||
More Information
Set Membership: |
|

LOC4343296
Genomic Map