Assay Details
| Gene Symbol: LOC4343510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789698.2 | XP_015645184.1 | 1 - 2 | 476 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4343510| Gene Symbol: LOC4343510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789698.2 | XP_015645184.1 | 1 - 2 | 476 | 87 | |
Set Membership: |
|